Learn More
Biomatik Corporation Mouse Nitric Oxide Synthase 2, Inducible (NOS2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06309
Encompass_Preferred
Mouse Nitric Oxide Synthase 2, Inducible (NOS2) ELISA Kit for detecting NOS2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29477. Gene ID: 18126. Target Synonyms: NOS2A; INOS; HEP-NOS; I-NOS; Hepatocytes Oxide Synthase; Peptidyl-cysteine S-nitrosylase NOS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3381