Learn More
Biomatik Corporation Rat Estrogen ELISA Kit
Supplier:  Biomatik Corporation EKF60790
Encompass_Preferred
Rat Estrogen ELISA Kit for detecting Estrogen in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: ESRRG; Estrogen-related receptor gamma; ERR3; NR3B3; ERR gamma-2; ERRG2; ERRgamma; Estrogen receptor-related protein 3; NR3B3; Nuclear receptor subfamily 3 group B member 3; KIAA0832. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8453