Learn More
Biomatik Corporation Rat Nitric Oxide Synthase 2, Inducible (NOS2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06310
Encompass_Preferred
Rat Nitric Oxide Synthase 2, Inducible (NOS2) ELISA Kit for detecting NOS2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06518. Gene ID: 24599. Target Synonyms: NOS2A; INOS; HEP-NOS; I-NOS; Hepatocytes Oxide Synthase; Peptidyl-cysteine S-nitrosylase NOS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3382