Learn More
Biomatik Corporation Human Nitric Oxide Synthase 3, Endothelial (NOS3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06312
Encompass_Preferred
The Human Nitric Oxide Synthase 3, Endothelial (NOS3) ELISA Kit allows for quantitative determination of Nitric Oxide Synthase 3, Endothelial in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29474. Cross Activity: No significant cross-reactivity or interference between Nitric Oxide Synthase 3, Endothelial and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3383