Learn More
Biomatik Corporation Human Nitric Oxide Synthase Trafficker (NOSTRIN) ELISA Kit, 96 tests
Catalog No. 501503386
Encompass_Preferred
The Human Nitric Oxide Synthase Trafficker (NOSTRIN) ELISA Kit allows for quantitative determination of Nitric Oxide Synthase Trafficker in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8IVI9. Cross Activity: No significant cross-reactivity or interference between Nitric Oxide Synthase Trafficker and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3386 Supplier Biomatik Corporation Supplier No. EKU06317
The Human Nitric Oxide Synthase Trafficker (NOSTRIN) ELISA Kit allows for quantitative determination of Nitric Oxide Synthase Trafficker in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8IVI9. Cross Activity: No significant cross-reactivity or interference between Nitric Oxide Synthase Trafficker and analogues was observed. Restriction: For Research Use Only.