Learn More
Biomatik Corporation Mouse Nodal Homolog (NODAL) ELISA Kit, 96 tests
Catalog No. 501503388
Encompass_Preferred
Mouse Nodal Homolog (NODAL) ELISA Kit for detecting NODAL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43021. Gene ID: 18119. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3388 Supplier Biomatik Corporation Supplier No. EKU06322
Mouse Nodal Homolog (NODAL) ELISA Kit for detecting NODAL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43021. Gene ID: 18119. Restrictions: For Research Use Only. Not for use in diagnostic procedures.