Learn More
Biomatik Corporation Noradrenaline (NE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06328
Encompass_Preferred
Noradrenaline (NE) ELISA Kit for detecting NE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 61.7-5,000pg/ml. Sensitivity: 24.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: NA; NAd; Norepinephrine; Norepi. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3389