Learn More
Biomatik Corporation Mouse Norepinephrine Transporter (NET) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06330
Encompass_Preferred
Mouse Norepinephrine Transporter (NET) ELISA Kit for detecting NET in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O55192. Gene ID: 20538. Target Synonyms: SLC6A2; NAT1; NET; NET1; Sodium-dependent noradrenaline transporter; Noradrenaline Transporter; Solute Carrier Family 6 member 2; Neurotransmitter transporter; Noradrenalin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3390