Learn More
Biomatik Corporation Monkey D2D (D-Dimer) ELISA Kit
Supplier:  Biomatik Corporation EKF60821
Encompass_Preferred
Monkey D2D (D-Dimer) ELISA Kit for detecting D2D in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P68109; G7P6F7; A0A2K5TY07. Target Synonyms: D2D; D-Dimer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8479