Learn More
Biomatik Corporation Human Notch Homolog 2 (NOTCH2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06332
Encompass_Preferred
The Human Notch Homolog 2 (NOTCH2) ELISA Kit allows for quantitative determination of Notch Homolog 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.781-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q04721. Cross Activity: No significant cross-reactivity or interference between Notch Homolog 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3392