Learn More
Biomatik Corporation Mouse N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit, 96 tests
Catalog No. 501503394
Encompass_Preferred
Mouse N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit for detecting NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40753. Gene ID: 18158. Target Synonyms: NT-Pro-BNP; ; N-BNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3394 Supplier Biomatik Corporation Supplier No. EKU06341
Mouse N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit for detecting NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40753. Gene ID: 18158. Target Synonyms: NT-Pro-BNP; ; N-BNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.