Learn More
Biomatik Corporation Rat N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06343
Encompass_Preferred
Rat N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit for detecting NT-ProBNP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13205. Gene ID: 25105. Target Synonyms: NT-Pro-BNP; ; N-BNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3396