Learn More
Biomatik Corporation Dog N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06345
Encompass_Preferred
The Dog N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit allows for quantitative determination of N-Terminal Pro-Brain Natriuretic Peptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 61.7-5000 pg/mL. Sensitivity: 23.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P16859. Cross Activity: No significant cross-reactivity or interference between N-Terminal Pro-Brain Natriuretic Peptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3397