Learn More
Biomatik Corporation Rat Nuclear Factor Kappa B (NFkB) ELISA Kit, 96 tests
Catalog No. 501503399
Encompass_Preferred
Rat Nuclear Factor Kappa B (NFkB) ELISA Kit for detecting NFkB in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63369. Gene ID: 81736. Target Synonyms: NF-KB1; EBP-1; KBF1; NF-Kappa-B; NFKB-P105; NFKB-P50; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 1; Nuclear factor NF-kappa-B p105 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3399 Supplier Biomatik Corporation Supplier No. EKU06354
Rat Nuclear Factor Kappa B (NFkB) ELISA Kit for detecting NFkB in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63369. Gene ID: 81736. Target Synonyms: NF-KB1; EBP-1; KBF1; NF-Kappa-B; NFKB-P105; NFKB-P50; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 1; Nuclear factor NF-kappa-B p105 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.