Learn More
Biomatik Corporation Mouse Nuclear Factor of Activated T-Cells, Cytoplasmic 1 (NFATC1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06357
Encompass_Preferred
Mouse Nuclear Factor Of Activated T-Cells, Cytoplasmic 1 (NFATC1) ELISA Kit for detecting NFATC1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88942. Target Synonyms: NF-ATC; NFAT2; NFATc; NFAT transcription complex cytosolic component. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3400