Learn More
Biomatik Corporation Mouse P III NP (N-Terminal Procollagen III Propeptide) ELISA Kit
Supplier:  Biomatik Corporation EKE61869
Encompass_Preferred
Mouse P III NP (N-Terminal Procollagen III Propeptide) ELISA Kit for detecting P III NP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.16-10ng/ml. Sensitivity: 0.1ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P08121. Target Synonyms: P3NP; N-Propeptide Of Type III Procollagen; Procollagen III Amino Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8521