Learn More
Biomatik Corporation Rat Nuclear Factor, Erythroid Derived 2 Like Protein 2 (NFE2L2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06361
Encompass_Preferred
Rat Nuclear Factor, Erythroid Derived 2 Like Protein 2 (NFE2L2) ELISA Kit for detecting NFE2L2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O54968. Gene ID: 83619. Target Synonyms: NRF2; HEBP1; NF-E2-Related Factor 2; Nuclear factor, erythroid derived 2, like 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3402