Learn More
Biomatik Corporation Rat IL-4 (Interleukin 4) ELISA Kit
Supplier:  Biomatik Corporation EKE61990
Encompass_Preferred
Rat IL-4 (Interleukin 4) ELISA Kit for detecting IL-4 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P20096. Target Synonyms: IL4; BCGF-1; BCGF1; BSF-1; BSF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8527