Learn More
Biomatik Corporation Mouse Occludin (OCLN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06383
Encompass_Preferred
Mouse Occludin (OCLN) ELISA Kit for detecting OCLN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.132ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61146. Gene ID: 18260. Target Synonyms: Tight Junction Protein Occludin TM4 Minus. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3410