Learn More
Biomatik Corporation Rat Occludin (OCLN) ELISA Kit, 96 tests
Catalog No. 501503411
Encompass_Preferred
Rat Occludin (OCLN) ELISA Kit for detecting OCLN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P6T5. Gene ID: 83497. Target Synonyms: Tight Junction Protein Occludin TM4 Minus. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3411 Supplier Biomatik Corporation Supplier No. EKU06384
Rat Occludin (OCLN) ELISA Kit for detecting OCLN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P6T5. Gene ID: 83497. Target Synonyms: Tight Junction Protein Occludin TM4 Minus. Restrictions: For Research Use Only. Not for use in diagnostic procedures.