Learn More
Biomatik Corporation Rat Orexin A (OXA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06399
Encompass_Preferred
Rat Orexin A (OXA) ELISA Kit for detecting OXA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.07pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: O55232. Gene ID: 25723. Target Synonyms: Hypocretin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3416