Learn More
Biomatik Corporation Human Orosomucoid 2 (ORM2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06409
Encompass_Preferred
The Human Orosomucoid 2 (ORM2) ELISA Kit allows for quantitative determination of Orosomucoid 2 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.27 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19652. Cross Activity: No significant cross-reactivity or interference between Orosomucoid 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3419