Learn More
Biomatik Corporation Mouse Osteocalcin (OC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06414
Encompass_Preferred
Mouse Osteocalcin (OC) ELISA Kit for detecting OC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P86546. Gene ID: 12096. Target Synonyms: BGLAP; OT; BGP; Bone Gla Protein; Bone Gamma-Carboxyglutamate Protein; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3422