Learn More
Biomatik Corporation Pig Osteocalcin (OC) ELISA Kit, 96 tests
Catalog No. 501503423
Encompass_Preferred
Pig Osteocalcin (OC) ELISA Kit for detecting OC in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8HYY9. Target Synonyms: BGLAP; OT; BGP; Bone Gla Protein; Bone Gamma-Carboxyglutamate Protein; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3423 Supplier Biomatik Corporation Supplier No. EKU06415
Pig Osteocalcin (OC) ELISA Kit for detecting OC in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8HYY9. Target Synonyms: BGLAP; OT; BGP; Bone Gla Protein; Bone Gamma-Carboxyglutamate Protein; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.