Learn More
Biomatik Corporation Human Osteonectin (ON) ELISA Kit, 96 tests
Catalog No. 501503428
Encompass_Preferred
The Human Osteonectin (ON) ELISA Kit allows for quantitative determination of Osteonectin in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.21 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09486. Cross Activity: No significant cross-reactivity or interference between Osteonectin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3428 Supplier Biomatik Corporation Supplier No. EKU06425
The Human Osteonectin (ON) ELISA Kit allows for quantitative determination of Osteonectin in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.21 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09486. Cross Activity: No significant cross-reactivity or interference between Osteonectin and analogues was observed. Restriction: For Research Use Only.