Learn More
Biomatik Corporation Human Myxovirus Resistance 1 (MX1) ELISA Kit
Supplier:  Biomatik Corporation EKL59226
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernatants and other biological fluids.. Species Reactivity: Human. Detection Range: 0.781-50ng/mL. Sensitivity: 0.27ng/mL. Format: 96-Well Plate. Assay Type: sandwich. Detection Method: sandwich. Assay Time: 3 hours. UniProt ID: P20591. Target Synonyms: IFI-78K,IFI78,MX,MxA,Interferon-Inducible Protein p78,Interferon-regulated resistance GTP-binding protein MxA. Restriction: For Research Use Only.
Catalog No. 50-228-8742