Learn More
Biomatik Corporation Rat Osteonectin (ON) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06427
Encompass_Preferred
Rat Osteonectin (ON) ELISA Kit for detecting ON in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.14ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16975. Gene ID: 24791. Target Synonyms: SPARC; BM-40; Secreted Protein,Acidic,Cysteine-Rich; Basement-membrane protein 40; Secreted protein acidic and rich in cysteine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3430