Learn More
Biomatik Corporation Cattle Osteopontin (OPN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06428
Encompass_Preferred
The Cattle Osteopontin (OPN) ELISA Kit allows for quantitative determination of Osteopontin in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P31096. Cross Activity: No significant cross-reactivity or interference between Osteopontin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3431