Learn More
Biomatik Corporation Human Neuropeptide Y (NPY) ELISA Kit
Catalog No. 502288752
Encompass_Preferred
Human Neuropeptide Y (NPY) ELISA Kit for detecting NPY in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.97pg/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 3h. Accession Number: P01303. Gene ID: 4852. Target Synonyms: NP-Y; PYY4; CPON; Pro-neuropeptide Y; Neuropeptide tyrosine; C-flanking peptide of NPY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-228-8752 Supplier Biomatik Corporation Supplier No. EKL59489
Add to Cart
Add to Cart
Human Neuropeptide Y (NPY) ELISA Kit for detecting NPY in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.97pg/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 3h. Accession Number: P01303. Gene ID: 4852. Target Synonyms: NP-Y; PYY4; CPON; Pro-neuropeptide Y; Neuropeptide tyrosine; C-flanking peptide of NPY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.