Learn More
Biomatik Corporation Dog Osteopontin (OPN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06429
Encompass_Preferred
The Dog Osteopontin (OPN) ELISA Kit allows for quantitative determination of Osteopontin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.134 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q3HTT3. Cross Activity: No significant cross-reactivity or interference between Osteopontin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3432