Learn More
Biomatik Corporation Human Osteoprotegerin (OPG) ELISA Kit, 96 tests
Catalog No. 501503435
Encompass_Preferred
The Human Osteoprotegerin (OPG) ELISA Kit allows for quantitative determination of Osteoprotegerin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O00300. Cross Activity: No significant cross-reactivity or interference between Osteoprotegerin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3435 Supplier Biomatik Corporation Supplier No. EKU06435
The Human Osteoprotegerin (OPG) ELISA Kit allows for quantitative determination of Osteoprotegerin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O00300. Cross Activity: No significant cross-reactivity or interference between Osteoprotegerin and analogues was observed. Restriction: For Research Use Only.