Learn More
Biomatik Corporation Rat Osteoprotegerin (OPG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06438
Encompass_Preferred
Rat Osteoprotegerin (OPG) ELISA Kit for detecting OPG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 15.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08727. Gene ID: 25341. Target Synonyms: TNFRSF11B; TNFRSF11-B; OCIF; OPG; TR1; Tumor Necrosis Factor Receptor Superfamily Member 11b; Osteoclastogenesis Inhibitory Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3437