Learn More
Biomatik Corporation Ovalbumin (OVA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06441
Encompass_Preferred
Ovalbumin (OVA) ELISA Kit for detecting OVA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 16-10,000pg/ml. Sensitivity: 6pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3439