Learn More
Biomatik Corporation Mouse Oxidized Low Density Lipoprotein (OxLDL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06445
Encompass_Preferred
Mouse Oxidized Low Density Lipoprotein (OxLDL) ELISA Kit for detecting OxLDL in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: OL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3441