Learn More
Biomatik Corporation Human Oxyntomodulin (OXM) ELISA Kit, 96 tests
Catalog No. 501503443
Encompass_Preferred
The Human Oxyntomodulin (OXM) ELISA Kit allows for quantitative determination of Oxyntomodulin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.17-500 ng/mL. Sensitivity: 2.73 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01275. Cross Activity: No significant cross-reactivity or interference between Oxyntomodulin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3443 Supplier Biomatik Corporation Supplier No. EKU06452
The Human Oxyntomodulin (OXM) ELISA Kit allows for quantitative determination of Oxyntomodulin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.17-500 ng/mL. Sensitivity: 2.73 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01275. Cross Activity: No significant cross-reactivity or interference between Oxyntomodulin and analogues was observed. Restriction: For Research Use Only.