Learn More
Biomatik Corporation Oxytocin (OT) ELISA Kit, 96 tests
Catalog No. 501503444
Encompass_Preferred
Oxytocin (OT) ELISA Kit for detecting OT in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.27pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Pitocin; Syntocinon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3444 Supplier Biomatik Corporation Supplier No. EKU06454
Oxytocin (OT) ELISA Kit for detecting OT in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.27pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Pitocin; Syntocinon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.