Learn More
Biomatik Corporation Human Oxytocin Receptor (OXTR) ELISA Kit, 96 tests
Catalog No. 501503445
Encompass_Preferred
The Human Oxytocin Receptor (OXTR) ELISA Kit allows for quantitative determination of Oxytocin Receptor in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P30559. Cross Activity: No significant cross-reactivity or interference between Oxytocin Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3445 Supplier Biomatik Corporation Supplier No. EKU06455
The Human Oxytocin Receptor (OXTR) ELISA Kit allows for quantitative determination of Oxytocin Receptor in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P30559. Cross Activity: No significant cross-reactivity or interference between Oxytocin Receptor and analogues was observed. Restriction: For Research Use Only.