Learn More
Biomatik Corporation Rat Oxytocin Receptor (OXTR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06456
Encompass_Preferred
Rat Oxytocin Receptor (OXTR) ELISA Kit for detecting OXTR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70536. Gene ID: 25342. Target Synonyms: OT-R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3446