Learn More
Biomatik Corporation Rat p53 Upregulated Modulator of Apoptosis (PUMA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06459
Encompass_Preferred
Rat p53 Upregulated Modulator Of Apoptosis (PUMA) ELISA Kit for detecting PUMA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q80ZG6. Gene ID: 317673. Target Synonyms: JFY1; BBC3; BCL2 Binding Component 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3447