Learn More
Biomatik Corporation Human Tissue Inhibitors Of Metalloproteinase 1 (TIMP1) ELISA Kit
Supplier:  Biomatik Corporation EKL61033
Encompass_Preferred
Human Tissue Inhibitors Of Metalloproteinase 1 (TIMP1) ELISA Kit for detecting TIMP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.1-5000pg/ml. Sensitivity: 37pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: P01033. Gene ID: 7076. Target Synonyms: EPO; CLGI; EPA; HCI; Erythroid Potentiating Activity; Collagenase Inhibitor; Metalloproteinase inhibitor 1; Fibroblast collagenase inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8795