Learn More
Biomatik Corporation Human Palate/Lung And Nasal Epithelium Associated Protein (PLUNC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06462
Encompass_Preferred
The Human Palate/Lung And Nasal Epithelium Associated Protein (PLUNC) ELISA Kit allows for quantitative determination of Palate/Lung And Nasal Epithelium Associated Protein in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NP55. Cross Activity: No significant cross-reactivity or interference between Palate/Lung And Nasal Epithelium Associated Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3448