Learn More
Biomatik Corporation Mouse Pancreatic Polypeptide (PP) ELISA Kit, 96 tests
Catalog No. 501503450
Encompass_Preferred
Mouse Pancreatic Polypeptide (PP) ELISA Kit for detecting PP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 9.16pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P10601. Gene ID: 19064. Target Synonyms: PPY; PNP; Pancreatic Polypeptide Y; Obinepitide; Pancreatic icosapeptide; Pancreatic prohormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3450 Supplier Biomatik Corporation Supplier No. EKU06467
Mouse Pancreatic Polypeptide (PP) ELISA Kit for detecting PP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 9.16pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P10601. Gene ID: 19064. Target Synonyms: PPY; PNP; Pancreatic Polypeptide Y; Obinepitide; Pancreatic icosapeptide; Pancreatic prohormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.