Learn More
Biomatik Corporation Human Von Willebrand Factor (vWF) ELISA Kit
Supplier:  Biomatik Corporation EKL61535
Encompass_Preferred
Human Von Willebrand Factor (vWF) ELISA Kit for detecting vWF in plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.53ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: P04275. Gene ID: 7450. Target Synonyms: F8VWF; VWD; von Willebrand antigen 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8812