Learn More
Biomatik Corporation Human Paraoxonase 1 (PON1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06474
Encompass_Preferred
The Human Paraoxonase 1 (PON1) ELISA Kit allows for quantitative determination of Paraoxonase 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.27 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P27169. Cross Activity: No significant cross-reactivity or interference between Paraoxonase 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3455