Learn More
Biomatik Corporation Rat Paraoxonase 1 (PON1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06476
Encompass_Preferred
Rat Paraoxonase 1 (PON1) ELISA Kit for detecting PON1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55159. Gene ID: 84024. Target Synonyms: ESA; PON; Esterase A; Serum paraoxonase/arylesterase 1; Aromatic esterase 1; A-esterase 1; Serum aryldialkylphosphatase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3457