Learn More
Biomatik Corporation Mouse Parathyroid Hormone (PTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06483
Encompass_Preferred
Mouse Parathyroid Hormone (PTH) ELISA Kit for detecting PTH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 9.88-800pg/ml. Sensitivity: 3.73pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9Z0L6. Gene ID: 19226. Target Synonyms: iPTH; Intact Parathyroid Hormone; Parathormone; Parathyrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3460