Learn More
Biomatik Corporation Rat Parathyroid Hormone (PTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06485
Encompass_Preferred
Rat Parathyroid Hormone (PTH) ELISA Kit for detecting PTH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 9.88-800pg/ml. Sensitivity: 4.39pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P04089. Gene ID: 24694. Target Synonyms: iPTH; Intact Parathyroid Hormone; Parathormone; Parathyrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3461