Learn More
Biomatik Corporation Cattle Parathyroid Hormone Related Protein (PTHrP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06488
Encompass_Preferred
The Cattle Parathyroid Hormone Related Protein (PTHrP) ELISA Kit allows for quantitative determination of Parathyroid Hormone Related Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P58073. Cross Activity: No significant cross-reactivity or interference between Parathyroid Hormone Related Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3463