Learn More
Biomatik Corporation Human Patatin Like Phospholipase Domain Containing Protein 3 (PNPLA3) ELISA Kit, 96 tests
Catalog No. 501503467
Encompass_Preferred
The Human Patatin Like Phospholipase Domain Containing Protein 3 (PNPLA3) ELISA Kit allows for quantitative determination of Patatin Like Phospholipase Domain Containing Protein 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NST1. Cross Activity: No significant cross-reactivity or interference between Patatin Like Phospholipase Domain Containing Protein 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3467 Supplier Biomatik Corporation Supplier No. EKU06497
The Human Patatin Like Phospholipase Domain Containing Protein 3 (PNPLA3) ELISA Kit allows for quantitative determination of Patatin Like Phospholipase Domain Containing Protein 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NST1. Cross Activity: No significant cross-reactivity or interference between Patatin Like Phospholipase Domain Containing Protein 3 and analogues was observed. Restriction: For Research Use Only.