Learn More
Biomatik Corporation Human Pentraxin 3, Long (PTX3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06503
Encompass_Preferred
The Human Pentraxin 3, Long (PTX3) ELISA Kit allows for quantitative determination of Pentraxin 3, Long in Serum, Plasma, Tissue homogenates, Cell lysates, Salivas and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.113 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P26022. Cross Activity: No significant cross-reactivity or interference between Pentraxin 3, Long and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3468